Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl propargyl ether, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
16314-18-4
C7H12O
112.17
MFCD07368651
QIGSPXPODKLGRK-UHFFFAOYSA-N
tert-butyl propargyl ether,3-tert-butoxy prop-1-yne,1-propyne,3-1,1-dimethylethoxy,1-propyne, 3-1,1-dimethylethoxy,3-tert-butoxyprop-1-yne,3-tert-butyloxy-1-propyne,tert-butylpropargylether
7172302
2-methyl-2-prop-2-ynoxypropane
CC(C)(C)OCC#C
16314-18-4
C7H12O
112.17
MFCD07368651
QIGSPXPODKLGRK-UHFFFAOYSA-N
tert-butyl propargyl ether,3-tert-butoxy prop-1-yne,1-propyne,3-1,1-dimethylethoxy,1-propyne, 3-1,1-dimethylethoxy,3-tert-butoxyprop-1-yne,3-tert-butyloxy-1-propyne,tert-butylpropargylether
7172302
2-methyl-2-prop-2-ynoxypropane
CC(C)(C)OCC#C
Boiling Point 119°C
Flash Point 20°C
Density 0.8350g/mL
Specific Gravity 0.835
Refractive Index 1.4170 to 1.419
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1g
Color Yellow
Assay Percent Range 96% min. (GC)
Linear Formula CH∽CCH2OC(CH3)3
Formula Weight 112.17
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material tert-Butyl propargyl ether
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger