Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ tert-Butyl (S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride, 98%

Click to view available options
Quantity:
250mg
1g
5g
77497-74-6
C14H19NO2
233.311
MFCD04114841
KRSUDBIZXKOKGA-LBPRGKRZSA-N
s-tert-butyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate,s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid t-butyl ester,tert-butyl 3s-1,2,3,4-tetrahydroisoquinoline-3-carboxylate,tert-butyl s-1,2,3,4-tetrahydro-isoquinoline-3-carboxylate,s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid tert-butyl ester,tic-obut,s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic ac,tert-butyl s-1,2,3,4-tetrahydroisoquinoline-3-carboxylate,tert.-butyl 3s-1,2,3,4-tetrahydroisoquinoline-3-carboxylate,s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid tert-butyl ester hydrochloride
9209323
tert-butyl (3S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate
CC(C)(C)OC(=O)C1CC2=CC=CC=C2CN1
77497-74-6
C14H19NO2
233.311
MFCD04114841
KRSUDBIZXKOKGA-LBPRGKRZSA-N
s-tert-butyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate,s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid t-butyl ester,tert-butyl 3s-1,2,3,4-tetrahydroisoquinoline-3-carboxylate,tert-butyl s-1,2,3,4-tetrahydro-isoquinoline-3-carboxylate,s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid tert-butyl ester,tic-obut,s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic ac,tert-butyl s-1,2,3,4-tetrahydroisoquinoline-3-carboxylate,tert.-butyl 3s-1,2,3,4-tetrahydroisoquinoline-3-carboxylate,s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid tert-butyl ester hydrochloride
9209323
tert-butyl (3S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate
CC(C)(C)OC(=O)C1CC2=CC=CC=C2CN1
Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Quantity 1g
Beilstein 5744730
Formula Weight 269.77
Physical Form Solid
Percent Purity 99%
Chemical Name or Material tert-Butyl (S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride

storageNote1 : 2° to 8°C