Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ tert-Butyl (S)-2-aminobutyrate hydrochloride, 95%

Click to view available options
Quantity:
1g
5g
25g
53956-05-1
C8H18ClNO2
195.687
MFCD00237697
OEPKETQBSXWOBJ-RGMNGODLSA-N
h-abu-otbu.hcl,s-tert-butyl 2-aminobutanoate hydrochloride,h-2-abu-otbu.hcl,h-2-abu-otbu hcl,tert-butyl 2s-2-aminobutanoate hydrochloride,s-2-aminobutyric acid t-butyl ester hydrochloride,butanoic acid, 2-amino-, 1,1-dimethylethyl ester, hydrochloride, s,2s-2-aminobutyric acid tert-butyl ester hydrochloride,h-abu-otbu hcl,h-2-abu-otbu hydrochloride
53628861
tert-butyl (2S)-2-aminobutanoate;hydrochloride
CCC(C(=O)OC(C)(C)C)N.Cl
53956-05-1
C8H18ClNO2
195.687
MFCD00237697
OEPKETQBSXWOBJ-RGMNGODLSA-N
h-abu-otbu.hcl,s-tert-butyl 2-aminobutanoate hydrochloride,h-2-abu-otbu.hcl,h-2-abu-otbu hcl,tert-butyl 2s-2-aminobutanoate hydrochloride,s-2-aminobutyric acid t-butyl ester hydrochloride,butanoic acid, 2-amino-, 1,1-dimethylethyl ester, hydrochloride, s,2s-2-aminobutyric acid tert-butyl ester hydrochloride,h-abu-otbu hcl,h-2-abu-otbu hydrochloride
53628861
tert-butyl (2S)-2-aminobutanoate;hydrochloride
CCC(C(=O)OC(C)(C)C)N.Cl
Quantity 5g
Beilstein 6935251
Formula Weight 195.69
Physical Form Solid
Percent Purity 95%
Chemical Name or Material tert-Butyl (S)-2-aminobutyrate hydrochloride

storageNote1 : 2° to 8°C