Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl (S)-(-)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, 95%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
102308-32-7
C11H19NO4
229.28
MFCD00209557
PNJXYVJNOCLJLJ-MRVPVSSYSA-N
s---3-boc-2,2-dimethyloxazolidine-4-carboxaldehyde,tert-butyl s---4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate,tert-butyl 4s-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate,unii-41aj913hqi,s-tert-butyl 4-formyl-2,2-dimethyloxazolidine-3-carboxylate,1,1-dimethylethyl-s-4-formyl-2,2-dimethyl-3-oxazolidine carboxylate,s---3-tert-butoxycarbonyl-4-formyl-2,2-dimethyl-1,3-oxazolidine,s-4-formyl-2,2-dimethyl-oxazolidine-3-carboxylic acid tert-butyl ester,3-oxazolidinecarboxylic acid, 4-formyl-2,2-dimethyl-, 1,1-dimethylethyl ester, 4s,s---3-boc-4-formyl-2,2-dimethyl-1,3-oxazolidine
179824
tert-butyl (4S)-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate
CC1(N(C(CO1)C=O)C(=O)OC(C)(C)C)C
102308-32-7
C11H19NO4
229.28
MFCD00209557
PNJXYVJNOCLJLJ-MRVPVSSYSA-N
s---3-boc-2,2-dimethyloxazolidine-4-carboxaldehyde,tert-butyl s---4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate,tert-butyl 4s-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate,unii-41aj913hqi,s-tert-butyl 4-formyl-2,2-dimethyloxazolidine-3-carboxylate,1,1-dimethylethyl-s-4-formyl-2,2-dimethyl-3-oxazolidine carboxylate,s---3-tert-butoxycarbonyl-4-formyl-2,2-dimethyl-1,3-oxazolidine,s-4-formyl-2,2-dimethyl-oxazolidine-3-carboxylic acid tert-butyl ester,3-oxazolidinecarboxylic acid, 4-formyl-2,2-dimethyl-, 1,1-dimethylethyl ester, 4s,s---3-boc-4-formyl-2,2-dimethyl-1,3-oxazolidine
179824
tert-butyl (4S)-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate
CC1(N(C(CO1)C=O)C(=O)OC(C)(C)C)C
Boiling Point 83°C to 88°C (1.0mmHg)
Density 1.0600g/mL
Flash Point 108°C
Packaging Glass Bottle
Specific Gravity 1.06
Specific Rotation -85° to -95° (20°C, 589nm) (c=1, CHCl3)
Color Colorless to Yellow
Refractive Index 1.4450 to 1.4490 (20°C, 589nm)
Quantity 250mg
Infrared Spectrum Authentic
Assay Percent Range 95%
Merck Index 15,4404
Formula Weight 229.28
Physical Form Oil
Percent Purity ≥94%
Chemical Name or Material tert-Butyl (S)-(-)-4-formyl-2, 2-dimethyl-3-oxazolidinecarboxylate
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning