Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ tert-Butyl (S)-5-oxopyrrolidine-2-carboxylate, 95%

Click to view available options
Quantity:
5g
25g
35418-16-7
C9H15NO3
185.223
MFCD06659481
QXGSPAGZWRTTOT-LURJTMIESA-N
s-tert-butyl 5-oxopyrrolidine-2-carboxylate,s-2-pyrrolidone-5-carboxylic acid t-butyl ester,tert-butyl 5-oxo-l-prolinate,l-pyroglutamic acid tert-butyl ester,tert-butyl s-2-pyrrolidone-5-carboxylate,tert-butyl l-pyroglutamate,tert-butyl 2s-5-oxopyrrolidine-2-carboxylate,s-5-oxo-pyrrolidine-2-carboxylic acid tert-butyl ester,l-pyroglutamicacidtert-butylester,tert-butyl s-5-oxopyrrolidine-2-carboxylate
10888648
tert-butyl (2S)-5-oxopyrrolidine-2-carboxylate
CC(C)(C)OC(=O)C1CCC(=O)N1
35418-16-7
C9H15NO3
185.223
MFCD06659481
QXGSPAGZWRTTOT-LURJTMIESA-N
s-tert-butyl 5-oxopyrrolidine-2-carboxylate,s-2-pyrrolidone-5-carboxylic acid t-butyl ester,tert-butyl 5-oxo-l-prolinate,l-pyroglutamic acid tert-butyl ester,tert-butyl s-2-pyrrolidone-5-carboxylate,tert-butyl l-pyroglutamate,tert-butyl 2s-5-oxopyrrolidine-2-carboxylate,s-5-oxo-pyrrolidine-2-carboxylic acid tert-butyl ester,l-pyroglutamicacidtert-butylester,tert-butyl s-5-oxopyrrolidine-2-carboxylate
10888648
tert-butyl (2S)-5-oxopyrrolidine-2-carboxylate
CC(C)(C)OC(=O)C1CCC(=O)N1
Color White
Quantity 25g
Beilstein 3590226
Formula Weight 185.22
Physical Form Powder
Percent Purity 95%
Chemical Name or Material tert-Butyl (S)-5-oxopyrrolidine-2-carboxylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning