Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl trimethylacetate, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
16474-43-4
C9H18O2
158.241
MFCD01861974
VXHFNALHLRWIIU-UHFFFAOYSA-N
tert-butyl pivalate,tert-butyl trimethylacetate,propanoic acid, 2,2-dimethyl, 1,1-dimethylethyl ester,di-tert-butylcarboxylic acid,2,2-dimethylpropanoic acid tert-butyl ester,2,2-dimethyl-propionic acid tert-butyl ester,propanoic acid, 2,2-dimethyl-, 1,1-dimethylethyl ester,propanoic acid,2,2-dimethyl-, 1,1-dimethylethyl ester
519272
tert-butyl 2,2-dimethylpropanoate
CC(C)(C)C(=O)OC(C)(C)C
16474-43-4
C9H18O2
158.241
MFCD01861974
VXHFNALHLRWIIU-UHFFFAOYSA-N
tert-butyl pivalate,tert-butyl trimethylacetate,propanoic acid, 2,2-dimethyl, 1,1-dimethylethyl ester,di-tert-butylcarboxylic acid,2,2-dimethylpropanoic acid tert-butyl ester,2,2-dimethyl-propionic acid tert-butyl ester,propanoic acid, 2,2-dimethyl-, 1,1-dimethylethyl ester,propanoic acid,2,2-dimethyl-, 1,1-dimethylethyl ester
519272
tert-butyl 2,2-dimethylpropanoate
CC(C)(C)C(=O)OC(C)(C)C
Packaging Glass bottle
Color Colorless
Density 0.8130g/mL
Refractive Index 1.3910 to 1.3930 (20°C, 589nm)
Infrared Spectrum Authentic
Quantity 5g
Assay Percent Range 98%
Linear Formula (CH3)3CCO2C(CH3)3
Formula Weight 158.24
Physical Form Liquid, Darkening During Storage
Percent Purity ≥97.5%
Chemical Name or Material tert-Butyl trimethylacetate
Show More Show Less