Alfa Aesar™ tert-Butylacetyl chloride, 98+%

Manufacturer: Alfa Aesar™ A1016222

Quantity 100g

 View more versions of this product

Catalog No. AAA1016222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7065-46-5
    Molecular Formula (CH3)3CCH2COCl
    Formula Weight 134.61
    MDL Number MFCD00000737
    UN Number UN2924
    Beilstein 1740421
    Synonym 3,3-Dimethylbutyryl chloride
    Chemical Name or Material tert-Butylacetyl chloride

    Specifications

    Formula Weight 134.61
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula (CH3)3CCH2COCl
    UN Number UN2924
    Synonym 3,3-Dimethylbutyryl chloride
    Health Hazard 1 Danger
    Boiling Point 128° to 130°C
    Flash Point 21°C (69°F)
    Recommended Storage 2°C to 8°C
    TSCA TSCA
    MDL Number MFCD00000737
    CAS 7065-46-5
    Beilstein 1740421
    Health Hazard 2 GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Odor Acrid
    Quantity 100g
    Chemical Name or Material tert-Butylacetyl chloride
    Assay ≥98%

    Description

    Applications
    tert-Butylacetyl chloride is used an acyl chloride compound for proteomics research. It is also used as intermediate for important acylation reagents agrochemical and pharmaceutical.

    Solubility
    Reacts with water.

    Notes
    Moisture Sensitive Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    2°C to 8°C

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit