Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ tert-Butylacetyl chloride, 98+%

Manufacturer:  Alfa Aesar™ A1016206

Quantity 5g

 View more versions of this product

Catalog No. AAA1016206

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
tert-Butylacetyl chloride is used an acyl chloride compound for proteomics research. It is also used as intermediate for important acylation reagents agrochemical and pharmaceutical.

Solubility
Reacts with water.

Notes
Moisture Sensitive Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Description & Specifications

Chemical Identifiers

7065-46-5
134.61
UN2924
3,3-Dimethylbutyryl chloride
(CH3)3CCH2COCl
MFCD00000737
1740421
tert-Butylacetyl chloride

Specifications

7065-46-5
134.61
UN2924
3,3-Dimethylbutyryl chloride
21°C (69°F)
5g
≥98%
(CH3)3CCH2COCl
MFCD00000737
1740421
128° to 130°C
Acrid
tert-Butylacetyl chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : TSCA

Recommended Storage : 2°C to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit