Alfa Aesar™ tert-Butylbenzene, 99%

Manufacturer: Alfa Aesar™ A12378AK

Quantity 250mL

 View more versions of this product

Catalog No. AAA12378AK

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 98-06-6
    Molecular Formula C10H14
    Formula Weight 134.22
    MDL Number MFCD00008816
    UN Number UN2709
    Beilstein 1421537
    Merck Index 14,1551
    Chemical Name or Material tert-Butylbenzene

    Specifications

    Formula Weight 134.22
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00008816
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTYL BENZENES
    Molecular Formula C10H14
    Merck Index 14,1551
    Health Hazard 2 GHS H Statement
    H226-H332-H315-H319-H335
    Flammable liquid and vapor.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Flash Point 34°C (93°F)
    Odor Characteristic
    Quantity 250mL
    CAS 98-06-6
    Beilstein 1421537
    Melting Point −58°C
    RTECS Number CY9120000
    UN Number UN2709
    Health Hazard 1 Warning
    Boiling Point 168° to 169°C
    TSCA TSCA
    Chemical Name or Material tert-Butylbenzene
    Assay 99%

    Description

    Applications
    tert-Butylbenzene is an useful intermediate for manufacturing agrochemicals and pharmaceuticals. tert-Butyl Toluene is used as a solvent for resins.

    Solubility
    Insoluble in water(0.03g/100 mL at 20°C) .

    Notes
    Keep container tightly closed in a cool, dry and well-ventilated place. Store away from strong oxidizing agents, acids.

    GHS H Statement
    H226-H332-H315-H319-H335
    Flammable liquid and vapor.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit