Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butylchlorodimethylsilane, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
18162-48-6
C6H15ClSi
150.73
MFCD00000501
BCNZYOJHNLTNEZ-UHFFFAOYSA-N
tert-butyldimethylsilyl chloride,tert-butyldimethylchlorosilane,t-butyldimethylchlorosilane,tert-butylchlorodimethylsilane,tbdms chloride,tert-butyl chloro dimethylsilane,chloro-tert-butyldimethylsilane,silane, chloro 1,1-dimethylethyl dimethyl,tbscl,t-butyldimethylsilyl chloride
28928
CHEBI:85071
tert-butyl-chloro-dimethylsilane
CC(C)(C)[Si](C)(C)Cl
18162-48-6
C6H15ClSi
150.73
MFCD00000501
BCNZYOJHNLTNEZ-UHFFFAOYSA-N
tert-butyldimethylsilyl chloride,tert-butyldimethylchlorosilane,t-butyldimethylchlorosilane,tert-butylchlorodimethylsilane,tbdms chloride,tert-butyl chloro dimethylsilane,chloro-tert-butyldimethylsilane,silane, chloro 1,1-dimethylethyl dimethyl,tbscl,t-butyldimethylsilyl chloride
28928
CHEBI:85071
tert-butyl-chloro-dimethylsilane
CC(C)(C)[Si](C)(C)Cl
Specific Gravity 0.87
Density 0.8700g/mL
Infrared Spectrum Authentic
Melting Point 85.0°C to 91.0°C
Boiling Point 125.0°C
Color Colorless to White
Packaging Glass bottle
Flash Point 22°C
Quantity 500g
Assay Percent Range 97.5% min. (GC)
Linear Formula (CH3)3CSi(CH3)2Cl
UN Number 2925
Fieser 04,176; 05,74; 06,78; 08,59; 09,77; 10,62; 13,87; 17,5
Merck Index 15, 1567
Solubility Information Solubility in water: reacts. Other solubilities: reacts with alcohols, organic acids and amines, soluble in ethers and hydrocarbons
Formula Weight 150.73
Physical Form Crystals
Percent Purity 98%
Chemical Name or Material tert-Butylchlorodimethylsilane
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Flammable solid.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or ha

GHS Signal Word: Danger

EINECSNumber : 242-042-4