Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butylchlorodiphenylsilane, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
58479-61-1
C16H19ClSi
274.86
MFCD00000497
MHYGQXWCZAYSLJ-UHFFFAOYSA-N
tert-butylchlorodiphenylsilane,tert-butyldiphenylchlorosilane,t-butyldiphenylchlorosilane,silane, chloro 1,1-dimethylethyl diphenyl,tert-butyl chloro diphenylsilane,t-butylchlorodiphenylsilane,tbdpscl,t-butyldiphenylsilyl chloride,tert-butyldiphenyl chlorosilane,unii-3beu48ui4e
94078
tert-butyl-chloro-diphenylsilane
CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl
58479-61-1
C16H19ClSi
274.86
MFCD00000497
MHYGQXWCZAYSLJ-UHFFFAOYSA-N
tert-butylchlorodiphenylsilane,tert-butyldiphenylchlorosilane,t-butyldiphenylchlorosilane,silane, chloro 1,1-dimethylethyl diphenyl,tert-butyl chloro diphenylsilane,t-butylchlorodiphenylsilane,tbdpscl,t-butyldiphenylsilyl chloride,tert-butyldiphenyl chlorosilane,unii-3beu48ui4e
94078
tert-butyl-chloro-diphenylsilane
CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl
Viscosity 16 mPa.s (25°C)
Boiling Point 135°C (0.7mmHg)
Flash Point 149°C
Specific Gravity 1.06
Refractive Index 1.5665 to 1.5685
Quantity 250g
Density 1.0600g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Color Brown or Colorless to Yellow
Assay Percent Range 97.5% min. (GC)
Linear Formula (CH3)3CSi(C6H5)2Cl
Merck Index 15, 1568
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in non-protic solvents,e.g. aliphatic,or aromatic (chlorinated) hydrocarbons
Formula Weight 274.86
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material tert-Butylchlorodiphenylsilane
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at

GHS Signal Word: Danger

EINECSNumber : 261-282-

Recommended Storage : May darken during storage