Alfa Aesar™ tert-Butylcyclohexane, 99+%

Manufacturer: Alfa Aesar™ B2412622

Quantity 100g

 View more versions of this product

Catalog No. AAB2412622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3178-22-1
    Molecular Formula C10H20
    Formula Weight 140.27
    MDL Number MFCD00001471
    UN Number UN3295
    Chemical Name or Material tert-Butylcyclohexane

    Specifications

    Formula Weight 140.27
    Molecular Formula C10H20
    MDL Number MFCD00001471
    CAS 3178-22-1
    UN Number UN3295
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapour.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 171°C
    Melting Point −41°C
    Flash Point 40°C (104°F)
    Quantity 100g
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
    RTECS Number GU9384375
    Chemical Name or Material tert-Butylcyclohexane
    Assay ≥99%

    Description

    Applications
    Used as pharmaceutical intermediate.

    Solubility
    Insoluble in water.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H226
    Flammable liquid and vapour.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit