Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(tert-Butyldimethylsilyl)acetylene, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
86318-61-8
C8H16Si
140.3
MFCD00191877
RTYNRTUKJVYEIE-UHFFFAOYSA-N
tert-butyldimethylsilyl acetylene,tert-butyldimethylsilylacetylene,tert-butyl ethynyl dimethylsilane,ethynyl-t-butyldimethylsilane,tert-butyldimethylsilyl ethyne,tert-butyl-ethynyl-dimethyl-silane,silane, 1,1-dimethylethyl ethynyldimethyl,t-butyldimethylsilylacetylene,tert-butyldimethylsilylethyne,tert-butylethynyldimethylsilane
2757281
tert-butyl-ethynyl-dimethylsilane
CC(C)(C)[Si](C)(C)C#C
86318-61-8
C8H16Si
140.3
MFCD00191877
RTYNRTUKJVYEIE-UHFFFAOYSA-N
tert-butyldimethylsilyl acetylene,tert-butyldimethylsilylacetylene,tert-butyl ethynyl dimethylsilane,ethynyl-t-butyldimethylsilane,tert-butyldimethylsilyl ethyne,tert-butyl-ethynyl-dimethyl-silane,silane, 1,1-dimethylethyl ethynyldimethyl,t-butyldimethylsilylacetylene,tert-butyldimethylsilylethyne,tert-butylethynyldimethylsilane
2757281
tert-butyl-ethynyl-dimethylsilane
CC(C)(C)[Si](C)(C)C#C
Boiling Point 116°C to 117°C
Flash Point 6°C
Specific Gravity 0.751
Density 0.7510g/mL
Color Colorless
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH∽CSi(CH3)2C(CH3)3
Formula Weight 140.3
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material (tert-Butyldimethylsilyl)acetylene
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger