Alfa Aesar™ tert-Butyldimethylsilyl trifluoromethanesulfonate, 98%

Manufacturer: Alfa Aesar™ A1217414

Quantity 25g

 View more versions of this product

Catalog No. AAA1217414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 69739-34-0
    Molecular Formula C7H15F3O3SSi
    Formula Weight 264.34
    MDL Number MFCD00000405
    UN Number UN2920
    Beilstein 2370068
    Synonym tert-Butyldimethylsilyl triflate; TBDMS-OTf
    Chemical Name or Material tert-Butyldimethylsilyl trifluoromethanesulfonate

    Specifications

    Formula Weight 264.34
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Synonym tert-Butyldimethylsilyl triflate; TBDMS-OTf
    Health Hazard 2 GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Molecular Formula C7H15F3O3SSi
    Beilstein 2370068
    Boiling Point 65° to 67°C (12mmHg)
    Quantity 25g
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
    CAS 69739-34-0
    UN Number UN2920
    Health Hazard 1 Danger
    Recommended Storage −30°C to −10°C
    MDL Number MFCD00000405
    Flash Point 36°C (96°F)
    Chemical Name or Material tert-Butyldimethylsilyl trifluoromethanesulfonate
    Assay 98%

    Description

    Applications
    tert-Butyldimethylsilyl trifluoromethanesulfonate is involved in the introduction of a bulky tert-butyl dimethylsilyl group onto a cis-bis(alkenyl)oxirane used in Cope rearrangement. It is associated with a thiolane and promotes the chalcogenide-Morita-Baylis-Hillman reaction. Further, it is used as a highly reactive silylating agent. In addition to this, it is used to prepare enol silyl ethers by reacting with ketones and lactones.

    Solubility
    Slightly miscible with chloroform.

    Notes
    Moisture sensitive. Store in a cool place. Incompatible with strong acids.

    GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.

    −30°C to −10°C

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit