Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ tert-Butylhydrazine hydrochloride, 98%

Click to view available options
Quantity:
25g
100g
500g
7400-27-3
C4H13ClN2
124.612
MFCD00012947
DDPWVABNMBRBFI-UHFFFAOYSA-N
tert-butylhydrazine hydrochloride,hydrazine, 1,1-dimethylethyl-, monohydrochloride,tert-butylhydrazinium chloride,tert-butylhydrazine monohydrochloride,t-butylhydrazine hydrochloride,hydrazine, 1,1-dimethylethyl-, hydrochloride 1:1,1,1-dimethylethyl hydrazine monohydrochloride,2-tert-butylhydrazinium chloride,t-butylhydrazine.hcl,t-bunhnh2.hcl
81889
tert-butylhydrazine;hydrochloride
CC(C)(C)NN.Cl
7400-27-3
C4H13ClN2
124.612
MFCD00012947
DDPWVABNMBRBFI-UHFFFAOYSA-N
tert-butylhydrazine hydrochloride,hydrazine, 1,1-dimethylethyl-, monohydrochloride,tert-butylhydrazinium chloride,tert-butylhydrazine monohydrochloride,t-butylhydrazine hydrochloride,hydrazine, 1,1-dimethylethyl-, hydrochloride 1:1,1,1-dimethylethyl hydrazine monohydrochloride,2-tert-butylhydrazinium chloride,t-butylhydrazine.hcl,t-bunhnh2.hcl
81889
tert-butylhydrazine;hydrochloride
CC(C)(C)NN.Cl
Applications
It is used in organic synthesis for aromatic nucleophilic N-N exchange reaction. tert-Butylhydrazine Hydrochloride is also used in the preparation of insect growth regulator. tert-Butylhydrazine hydrochloride was used in the synthesis of diphosphinohydrazines, 5-amino-1-tert-butyl-3-(p-chlorophenyl)-4-cyanopyrazole and 1,3,4,5-tetrasubstituted pyrazole derivatives.

Solubility
Solubility in water (almost transparency). Soluble in DMSO and methanol.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity and water.
Melting Point ~192°C (decomposition)
Quantity 100g
Beilstein 3651224
Formula Weight 124.61
Percent Purity 98%
Chemical Name or Material tert-Butylhydrazine hydrochloride

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : MV2841000

TSCA : TSCA