Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butylhydrazine hydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
7400-27-3
C4H12N2·HCl
124.61
MFCD00012947
DDPWVABNMBRBFI-UHFFFAOYSA-N
tert-butylhydrazine hydrochloride,hydrazine, 1,1-dimethylethyl-, monohydrochloride,tert-butylhydrazinium chloride,tert-butylhydrazine monohydrochloride,t-butylhydrazine hydrochloride,hydrazine, 1,1-dimethylethyl-, hydrochloride 1:1,1,1-dimethylethyl hydrazine monohydrochloride,2-tert-butylhydrazinium chloride,t-butylhydrazine.hcl,t-bunhnh2.hcl
81889
tert-butylhydrazine;hydrochloride
CC(C)(C)NN.Cl
7400-27-3
C4H12N2·HCl
124.61
MFCD00012947
DDPWVABNMBRBFI-UHFFFAOYSA-N
tert-butylhydrazine hydrochloride,hydrazine, 1,1-dimethylethyl-, monohydrochloride,tert-butylhydrazinium chloride,tert-butylhydrazine monohydrochloride,t-butylhydrazine hydrochloride,hydrazine, 1,1-dimethylethyl-, hydrochloride 1:1,1,1-dimethylethyl hydrazine monohydrochloride,2-tert-butylhydrazinium chloride,t-butylhydrazine.hcl,t-bunhnh2.hcl
81889
tert-butylhydrazine;hydrochloride
CC(C)(C)NN.Cl
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 190°C to 194°C
Color White
Quantity 100g
Assay Percent Range 98%
Linear Formula (CH3)3CNHNH2·HCl
Beilstein 04, III, 1734
Fieser 12,87
Formula Weight 124.61
Physical Form Crystals or Crystalline Powder
Percent Purity 98%
Chemical Name or Material tert-Butylhydrazine hydrochloride, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 231-001-6

RTECSNumber : MV2841000

TSCA : TSCA