Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butylhydroquinone 98.0+%, TCI America™
SDP

Catalog No. p-7118700
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 1948-33-0
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00002344
InChI Key BGNXCDMCOKJUMV-UHFFFAOYSA-N
Synonym tert-butylhydroquinone, tbhq, 2-tert-butylhydroquinone, t-butylhydroquinone, mtbhq, t-butyl hydroquinone, 2-tert-butyl-1,4-benzenediol, 2-t-butylhydroquinone, mono-tert-butylhydroquinone, sustane
PubChem CID 16043
ChEBI CHEBI:78886
IUPAC Name 2-tert-butylbenzene-1,4-diol
SMILES CC(C)(C)C1=C(C=CC(=C1)O)O

Specifications

Melting Point 127°C
Boiling Point 288°C
Quantity 25 g
UN Number 3077
Formula Weight 166.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butylhydroquinone
EINECSNumber (3)-0549
RTECSNumber MX4375000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.