Learn More
Alfa Aesar™ tert-Butyllithium, nominally 1.5M in n-pentane
Manufacturer:  Alfa Aesar™ 44125-JQ
Quantity 0.1 mole
 View more versions of this product
Catalog No. AA44125JQ
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Chemical Identifiers
594-19-4
64.06
UN3394
tert-Butyllithium
(CH3)3CLi
MFCD00008795
3587204
Specifications
Nominally 1.5M in n-pentane
0.1 mole
Colorless to pale yellow
Air & Moisture Sensitive
(CH3)3CLi
MFCD00008795
3587204
594-19-4
0.652g/mL
−49°C (−56°F)
Liquid
64.06
UN3394
tert-Butyllithium
Safety and Handling
GHS H Statement
H250-H260-H252-H304-H314-H336
GHS P Statement
P210-P222-P231+P232-P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Danger
DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
TSCA : TSCA