Alfa Aesar™ tert-Butyllithium, nominally 1.5M in n-pentane, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer: Alfa Aesar™ 41251AD

Quantity 50mL

 View more versions of this product

Catalog No. AA41251AD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 594-19-4
    Linear Formula (CH3)3CLi
    Formula Weight 64.06
    MDL Number MFCD00008795
    UN Number UN3394
    Beilstein 3587204
    Chemical Name or Material tert-Butyllithium

    Specifications

    Formula Weight 64.06
    Health Hazard 2 GHS H Statement
    H250-H260-H252-H304-H314-H336
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Self-heating in large quantities
    may catch fire.
    May be fatal if swallowed and enters airways.
    Cau
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    Quantity 50mL
    Health Hazard 1 Danger
    Storage Note 1
    • Packaged under Argon
    DOT Information Transport Hazard Class: 4.2; Packaging Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
    TSCA TSCA
    Beilstein 3587204
    Density 0.652g/mL
    Color Colorless to pale yellow
    Sensitivity Air & Moisture Sensitive
    MDL Number MFCD00008795
    CAS 594-19-4
    UN Number UN3394
    Packaging ChemSeal™ bottles
    Concentration Nominally 1.5M in n-pentane
    Linear Formula (CH3)3CLi
    Chemical Name or Material tert-Butyllithium

    GHS H Statement
    H250-H260-H252-H304-H314-H336
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Self-heating in large quantities
    may catch fire.
    May be fatal if swallowed and enters airways.
    Cau

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit