Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ tert-Butylmagnesium chloride, 1M in THF

Click to view available options
Quantity:
100mL
677-22-5
C4H9ClMg
116.871
MFCD00000465
CQRPUKWAZPZXTO-UHFFFAOYSA-M
tert-butylmagnesium chloride,tert-butylmagnesiumchloride,t-butylmagnesium chloride,t-butyl magnesium chloride,tert-butyl chloro magnesium,magnesium, chloro 1,1-dimethylethyl,tert-butylmagnesium chloride solution, 1.0 m in thf,tbumgcl,tert-bumgcl,t-bumgcl
2724198
magnesium;2-methylpropane;chloride
C[C-](C)C.[Mg+2].[Cl-]
677-22-5
C4H9ClMg
116.871
MFCD00000465
CQRPUKWAZPZXTO-UHFFFAOYSA-M
tert-butylmagnesium chloride,tert-butylmagnesiumchloride,t-butylmagnesium chloride,t-butyl magnesium chloride,tert-butyl chloro magnesium,magnesium, chloro 1,1-dimethylethyl,tert-butylmagnesium chloride solution, 1.0 m in thf,tbumgcl,tert-bumgcl,t-bumgcl
2724198
magnesium;2-methylpropane;chloride
C[C-](C)C.[Mg+2].[Cl-]
Sensitivity Air & Moisture Sensitive
Quantity 100mL
UN Number UN3399
Concentration or Composition (by Analyte or Components) 1M in THF
Physical Form Liquid
Chemical Name or Material tert-Butylmagnesium chloride

GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.

GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packaging Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE