Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butylperoxy 2-ethylhexyl carbonate, 95%, Acros Organics

Click to view available options
Quantity:
10mL
100mL
34443-12-4
C13H26O4
246.347
BRQMAAFGEXNUOL-UHFFFAOYSA-N
tert-butylperoxy 2-ethylhexyl carbonate,carbonoperoxoic acid, oo-1,1-dimethylethyl o-2-ethylhexyl ester,trigonox 117,tert-butylperoxy 2-ethylhexyl monocarbonate,oo-t-butyl o-2-ethylhexyl monoperoxycarbonate,oo-tert-butyl o-2-ethylhexyl peroxycarbonate,dsstox_cid_24921,dsstox_rid_80587,dsstox_gsid_44921,tert-butoxy 2-ethylhexyl carbonate
61931
2-ethylhexyl (2-methylpropan-2-yl)oxy carbonate
CCCCC(CC)COC(=O)OOC(C)(C)C
34443-12-4
C13H26O4
246.347
BRQMAAFGEXNUOL-UHFFFAOYSA-N
tert-butylperoxy 2-ethylhexyl carbonate,carbonoperoxoic acid, oo-1,1-dimethylethyl o-2-ethylhexyl ester,trigonox 117,tert-butylperoxy 2-ethylhexyl monocarbonate,oo-t-butyl o-2-ethylhexyl monoperoxycarbonate,oo-tert-butyl o-2-ethylhexyl peroxycarbonate,dsstox_cid_24921,dsstox_rid_80587,dsstox_gsid_44921,tert-butoxy 2-ethylhexyl carbonate
61931
2-ethylhexyl (2-methylpropan-2-yl)oxy carbonate
CCCCC(CC)COC(=O)OOC(C)(C)C
Radical initiators
Quantity 10mL
Packaging Plastic bottle
Assay Percent Range 95%
Formula Weight 246.35
Chemical Name or Material tert-Butylperoxy 2-ethylhexyl carbonate