Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
tert-Butylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed in cross couplig reactions.
Solubility
Reacts with water.
Notes
Handle under dry protective gas. Air sensitive. Store away from strong bases and oxidizing agents, heat, acids and wate
It is employed in cross couplig reactions.
Solubility
Reacts with water.
Notes
Handle under dry protective gas. Air sensitive. Store away from strong bases and oxidizing agents, heat, acids and wate
Specifications
Density | 0.966 |
Linear Formula | (CH3)3CZnBr |
Quantity | 50 mL |
UN Number | UN3399 |
Sensitivity | Air sensitive; May form a precipitate |
Solubility Information | Reacts with water. |
Formula Weight | 202.4 |
Chemical Name or Material | tert-Butylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles |
Safety and Handling
Hazard Category | H225-H260-H302-H314-H335-H336-H351 |
Hazard Statement | GHS H Statement H225-H261-H314-H351-H335 Highly flammable liquid and vapor. In contact with water releases flammable gas. Causes severe skin burns and eye damage. Suspected of causing cancer. May cause respiratory irritation. |
Precautionary Statement | P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE |
TSCA | No |
Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction