Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ tert-Pentylamine, 98%

Click to view available options
Quantity:
25g
100g
594-39-8
C5H13N
87.166
MFCD00008056
GELMWIVBBPAMIO-UHFFFAOYSA-N
tert-amylamine,tert-pentylamine,2-butanamine, 2-methyl,t-amylamine,1,1-dimethylpropylamine,2-methyl-2-butylamine,2-amino-2-methylbutane,2-methyl-butan-2-amine,1,1-dimethyl-1-propylamine,t-pentylamine
68986
CHEBI:84236
2-methylbutan-2-amine
CCC(C)(C)N
594-39-8
C5H13N
87.166
MFCD00008056
GELMWIVBBPAMIO-UHFFFAOYSA-N
tert-amylamine,tert-pentylamine,2-butanamine, 2-methyl,t-amylamine,1,1-dimethylpropylamine,2-methyl-2-butylamine,2-amino-2-methylbutane,2-methyl-butan-2-amine,1,1-dimethyl-1-propylamine,t-pentylamine
68986
CHEBI:84236
2-methylbutan-2-amine
CCC(C)(C)N
Flash Point −1°C (30°F)
Odor Amine-like
Boiling Point 76°C to 78°C
Quantity 25g
UN Number UN1106
Formula Weight 87.17
Percent Purity 98%
Chemical Name or Material tert-Pentylamine

GHS H Statement
H225-H314-H318-H302-H312-H332
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYLAMINES

TSCA : TSCA