Terumo™ Capiject™ Capillary Blood Collection Tubes Available on GSA/VA Contract for Federal Government customers only.

Single-piece, tapered design for easy collection and handling

Manufacturer: Terumo™ TMQK

 View more versions of this product

Catalog No. 22-042607

  Check Availability
Add to Cart

    Specifications

    Product Type Single-piece, tapered tube
    Capacity (Metric) 500μL
    Applications Most procedures requiring whole blood or plasma
    Closure Included Yes
    Additive Potassium EDTA (0.78mg of dipotassium EDTA)
    Color Lavender

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit