Terumo™ Capiject™ Capillary Blood Collection Tubes Available on GSA/VA Contract for Federal Government customers only.
Single-piece, tapered design for easy collection and handling
Manufacturer: Terumo™ TM
 View more versions of this product
Catalog No. 22-225516
  Check Availability
Add to Cart
    Specifications
    Product Type Single-piece, tapered tube
    Capacity (Metric) 500μL
    Additive Plain (no additives)
    Color Red
    Applications Used when a clotted sample is required; especially useful for blood banking or other laboratory procedures requiring serum
    Closure Included Yes
  • Description & Specifications
Related Products
Product Content Feedback