Terumo™ Capiject™ Capillary Blood Collection Tubes Available on GSA/VA Contract for Federal Government customers only.

Single-piece, tapered design for easy collection and handling

Manufacturer: Terumo™ TMG

 View more versions of this product

Catalog No. 22-225524

  Check Availability
Add to Cart

    Specifications

    Capacity (Metric) 500μL
    Applications Forms a barrier between serum and cells when the tube is centrifuged
    Additive Gel Barrier (inert gel) with clot activator (silica particles)
    Color Cherry Red
    Closure Included Yes
    Product Type Single-piece, tapered tube

    • Extra-wide, built-in collection lip for enhanced specimen flow
    • Color-coded fill lines and caps for volume accuracy and easy identification
    • Tube nests in ridged cap for convenient stand-alone placement
    • Lot number and expiration date printed on every tube
    • Silicone-coated anticoagulant tubes designed to reduce clotting
    • Olefin-based gel for clean separations
    • Certified for lead testing
    • Latex-free

    Tube Functions:

    • Potassium EDTA: Provides a ratio of anti-coagulant to blood that is optimal for most procedures requiring whole blood or plasma
    • Lithium Heparin: Useful for many chemistry procedures that require heparinized whole blood or plasma
    • Sodium Fluoride/Potassium Oxalate: Optimal for procedures where glycolysis inhibitions are important
    • Gel Barrier: Forms a barrier between serum and cells when the tube is centrifuged
    • Amber-Colored Tube: Useful for preserving light-sensitive sample integrity
    • Plain Tube: Used when a clotted sample is required; especially useful for blood banking or other laboratory procedures requiring serum

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit