Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Terumo™ Capiject™ Capillary Blood Collection Tubes
Single-piece, tapered design for easy collection and handling
Manufacturer:  Terumo™ TMLHG
 View more versions of this product
Catalog No. 22-256536
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Yes
500μL
Lithium Heparin
Single-piece, tapered tube
Useful for many chemistry procedures that require heparinized whole blood or plasma
Mint Green