Learn More
Terumo™ Capiject™ Capillary Blood Collection Tubes
Single-piece, tapered design for easy collection and handling
Manufacturer:  Terumo™ 3TMLHGA
 View more versions of this product
Catalog No. 22-225-578
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Yes
500μL
Lithium Heparin (12.5) with Gel Barrier (inert gel)
Single-piece, tapered tube
Useful for many chemistry procedures that require heparinized whole blood or plasma; Amber color Useful for preserving light-sensitive sample integrity
Mint Green, Amber tube