Learn More
Terumo™ SurGuard™3 Safety Hypodermic Needles, 23G
With three ways to activate (finger, thumb and hard surface) and a broad range of sizes, SurGuard™3 is the industry's most adaptable safety needle.
Supplier: Terumo™ SG32325
Description
- 3 modes of activation-thumb, finger, or hard surface
- Separate finger/thumb grips with ridges and nonslip surfaces
- Hub and needle locks for security post activation
- Broad range of sizes
- Firm injection feel
- Sharpest needles among major brands tested; 10% sharper than similar needles from Becton, Dickinson and Company
- More comfortable injections for patients
Specifications
23 G | |
1 in. | |
Sharp |
Yes | |
Hypodermic Safety Needle | |
Sterile |
Safety and Handling
ShelfLife : 5 Years