Learn More
Terumo™ VENOJECT™ II Evacuated Tube Holders
Reusable polypropylene holders for all tubes regardless of manufacturer
Manufacturer:  Terumo™ P1316R
Catalog No. 22-293057
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Shape and flange allow for easier, safer handling
  • Guideline facilitates positioning the tube before venipuncture
  • Multi-sample Luer adapter is sterile and disposable adapters accommodate Luer devices
  • Modify all Venoject II holders to accommodate Luer hub hypodermic needles for multiple use of evacuated tubes
Specifications
Specifications
Evacuated Tube Holder
Shape and flange allow for easier, safer handling
Multi sample luer adapter and Diversion Blood Sampling Arm™ and Blood Sampling Arm™ Blood bags
Polypropylene