Learn More
TES, 99%, Thermo Scientific Chemicals
Description
TES plays an important role in production of pH buffers for a biological system. It is used to make zwitterionic buffer solutions in the pH range 6.8-8.2. It is used in molecular biology, diagnosis, cell culture, pharmacy, agrochemical and pharmaceuticals. It is also used in the study of succinate oxidation and one of the components of test yolk buffer medium used in refrigeration of semen and transport.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TES plays an important role in production of pH buffers for a biological system. It is used to make zwitterionic buffer solutions in the pH range 6.8-8.2. It is used in molecular biology, diagnosis, cell culture, pharmacy, agrochemical and pharmaceuticals. It is also used in the study of succinate oxidation and one of the components of test yolk buffer medium used in refrigeration of semen and transport.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | ∼225°C (decomposition) |
Odor | Odorless |
Linear Formula | (HOCH2)3CNHCH2CH2SO3H |
Quantity | 50 g |
Beilstein | 1957061 |
Merck Index | 14,9176 |
Solubility Information | Soluble in water. |
Formula Weight | 229.25 |
Percent Purity | 99% |
Chemical Name or Material | TES |
Safety and Handling
EINECSNumber | 230-906-3 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.