Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TES, CellPURE™, Fisher BioReagents

Click to view available options
Quantity:
100g
Packaging:
Poly Bottle
7365-44-8
C6H15NO6S
229.247
JOCBASBOOFNAJA-UHFFFAOYSA-N
N-[Tris(hydroxymethyl)methyl]-2-aminoethanesulfonic acid, TES free acid, 2-[(2-Hydroxy-1, 1-bis(hydroxymethyl)ethyl)amino]ethanesulfonic acid
81831
CHEBI:44356
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonic acid
C(CS(=O)(=O)O)NC(CO)(CO)CO
7365-44-8
C6H15NO6S
229.247
JOCBASBOOFNAJA-UHFFFAOYSA-N
N-[Tris(hydroxymethyl)methyl]-2-aminoethanesulfonic acid, TES free acid, 2-[(2-Hydroxy-1, 1-bis(hydroxymethyl)ethyl)amino]ethanesulfonic acid
81831
CHEBI:44356
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonic acid
C(CS(=O)(=O)O)NC(CO)(CO)CO
Purity 99% min.
Absorbance 0.1 AU max. at 290nm
Additional Information Vapor Pressure: Negligible
Cell Culture Test Pass Test
Assay Percent Range ≥99 %
Endotoxin 0.1 EU/mg max.
Heavy Metals (as Pb) 5ppm max.
Formula Weight 229.25
Physical Form Powder/Solid
Grade CellPURE™
Infrared Spectrum Conforms to reference
Packaging Poly Bottle
Standard Plate Count 100 CFU/g max.
Water 0.2% max.
Vapor Density 7.9g/mol
Decomposition Information >220°C
RNase, DNase, Nickase, and Protease Not Detected Detected/Not Detect.
Melting Point 223°C
pH 4.0 to 4.8
Color White
Quantity 100g
Chemical Name or Material TES, Cell culture tested
Show More Show Less

WARNING!

Emergency Overview
Irritating to eyes, respiratory system and skin. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:2
Flammability:1
Instability:0