Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TESK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5121550

This item is not returnable.
View return policy
Description
TESK1 Polyclonal antibody specifically detects TESK1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
TESK1 | |
Polyclonal | |
Unconjugated | |
TESK1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
O70146, Q15569, Q63572 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.57 mg/mL | |
PBS with 50% glycerol and 0.05% Proclin 300 | |
PBS with 50% glycerol and 0.05% Proclin 300 | |
TESK1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title