Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRSS41 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548393

Catalog No. PA548393


Add to cart

Specifications

Specifications

PRSS41
Polyclonal
PBS with 0.09% sodium azide
Q7RTY9
Putative serine protease 41, 3421-, Testis serine protease 1, TESSP-1, PRSS41, TESSP1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
360226
Western Blot
Unconjugated
PRSS41
Liquid
PRSS41
KLH conjugated synthetic peptide between 106-133 amino acids from the Central region of human TESSP1.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit