Learn More
GSC Go Science Crazy Test Tube Brushes Products for Science Education
Brush and clean test tubes in laboratory or classroom settings. GSC International Test Tube Brushes employ soft, natural bristles to avoid scratching and galvanized handles to prevent rust.
Manufacturer:  GSC Go Science Crazy 41027010
 View more versions of this product
Catalog No. S13721
Add to cart
Alert:
Intended for ages 13 and up.
Description
Description
  • Great for cleaning out test tubes in laboratory or classroom settings
  • Soft, natural bristles are designed to clean tubes without scratching
  • Galvanized handles prevent rusting over repeated use
  • Available in a variety of sizes to fit different size tubes
Specifications
Specifications
Education
35mm
Cleaning test tubes
75mm
Soft natural bristles
240mm
Galvanized steel