Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Testican 3/SPOCK3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159703
Description
Testican 3/SPOCK3 Polyclonal specifically detects Testican 3/SPOCK3 in Human samples. It is validated for Western Blot.Specifications
Testican 3/SPOCK3 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Q9BQ16-1 | |
SPOCK3 | |
Synthetic peptides corresponding to SPOCK3(sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3) The peptide sequence was selected from the middle region of SPOCK3. Peptide sequence VDRYGNEVMGSRINGVADCAIDFEISGDFASGDFHEWTDDEDDEDDIMND The peptide sequence for this immunogen was taken from within the described region. | |
Affinity Purified | |
RUO | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
CWCV, and Kazal-like domains proteoglycan 3, HSAJ1454, sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3, TES-3, testican 3, TICN3 | |
Rabbit | |
49 kDa | |
100 μL | |
Signal Transduction | |
50859 | |
Human, Mouse, Rat, Porcine, Canine, Guinea Pig, Rabbit | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title