Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Testican 3/SPOCK3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | Testican 3/SPOCK3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP159703
|
Novus Biologicals
NBP159703 |
100 μL |
Each of 1 for $436.00
|
|
Description
Testican 3/SPOCK3 Polyclonal specifically detects Testican 3/SPOCK3 in Human samples. It is validated for Western Blot.Specifications
Testican 3/SPOCK3 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
CWCV, and Kazal-like domains proteoglycan 3, HSAJ1454, sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3, TES-3, testican 3, TICN3 | |
SPOCK3 | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
Q9BQ16-1 | |
50859 | |
IgG | |
Affinity Purified | |
49 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title