Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Testosterone Mouse anti-Nonspecies specific, Clone: BGN/06/902, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | Testosterone |
---|---|
Clone | BGN/06/902 |
Dilution | ELISA 1:100-1:2000 |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10062942
|
Novus Biologicals
NB10062942 |
0.1 mg |
Each for $436.00
|
N/A |
NB017521
|
Novus Biologicals
NB100629420.025MG |
0.025 mg |
Each for $176.85
|
N/A |
Description
Testosterone Monoclonal specifically detects Testosterone in Non-species specific samples. It is validated for ELISA.Specifications
Testosterone | |
ELISA 1:100-1:2000 | |
Monoclonal | |
Mouse | |
Nonspecies specific | |
IgG3 | |
Affinity Purified | |
This product is specific for Testosterone. It has shown the following reactivities. Testosterone 100% 19 Nortestosterone 15% Dihydrotestosterone <1% Testosterone promotes the development and maintenance of male secondary sex characteristics and is produced primarily in the testes. It is also produced in small amounts by the adrenal glands, ovaries and placenta. Synthetic derivatives of testosterone are used therapeutically in the treatment of breast cancer and for hormone replacement therapy. |
BGN/06/902 | |
ELISA | |
Unconjugated | |
RUO | |
Testosterone-11-HS-BSA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title