Learn More
Testosterone Mouse, Clone: 4E1G2, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB14425
Catalog No. 89-034-508
Add to cart
Description
Description
Mouse monoclonal antibody raised against human testosterone.
  • High specificity
  • High reactivity
Specifications
Specifications
Testosterone
4E1G2
Mouse monoclonal antibody raised against human testosterone.
In 10 mM PBS (0.05% BSA and 0.05% azide).
Mouse
IgG1, kappa
100 ug
Store at 4°C.
Monoclonal
ELISA, Immunohistochemistry (PFA fixed), Radioimmune Assays
Unconjugated
ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) Radioimmunoassay The optimal working dilution should be determined by the end user.
Liquid
Testosterone-3-carboxy-methyl-oxyme conjugated to BSA.
Protein A/G purification
RUO
Primary
Reacts with testosterone 100% 11-beta-hydroxy testosterone 3.3% 17-alpha-methyl testosterone <0.1% 5-alpha DHT 0.8% estradiol <0.1% progesterone <0.1%
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)