Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TET1 Rabbit anti-Mouse, DyLight™ 350, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178966UV

Catalog No. NBP178966UV


Add to cart

Description

Description

TET1 Polyclonal antibody specifically detects TET1 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

TET1
DyLight™ 350
50 mM sodium borate with 0.05% sodium azide
bA119F7.1, CXXC finger 6, CXXC6, CXXC6FLJ10839, FLJ41442, KIAA1676CXXC-type zinc finger protein 6, LCX, LCXEC 1.14.11.n2, Leukemia-associated protein with a CXXC domain, methylcytosine dioxygenase TET1, Ten-eleven translocation 1 gene protein, ten-eleven translocation-1, tet oncogene 1
Rabbit
IgG
0.1 mL
Primary
80312.0
Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry, Immunohistochemistry-Paraffin
Affinity Purified, Conjugated
TET1
A synthetic peptide made to an internal portion of the human TET1 protein (between residues 1700 to 1800). [UniProt Q8NFU7]
Immunogen affinity purified
Store at 4°C in the dark.
Polyclonal
Mouse
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit