Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TET1 Mouse anti-Human, Clone: OTI8G6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526527

Catalog No. PIMA526527


Add to cart

Description

Description

TET1 (tet oncogene 1), also known as LCX or CXXC6, is a 2,136 amino acid protein that localizes to the nucleus and contains one CXXC-type zinc finger. Expressed in adult ovary, thymus and skeletal muscle and also present in fetal lung, heart and brain, TET1 is thought to play a role in the development of fetal organs and may also be involvement in the pathoegenesis and metastasis of acute myeloid leukemia (AML). The gene encoding TET1 maps to human chromosome 10, which houses over 1,200 genes and comprises nearly 4.5% of the human genome. Defects in some of the genes that map to chromosome 10 are associated with Charcot-Marie Tooth disease, Jackson-Weiss syndrome, Usher syndrome, nonsyndromatic deafness, Wolman ft.s syndrome, Cowden syndrome, multiple endocrine neoplasia type 2 and porphyria.
Specifications

Specifications

TET1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8NFU7
RP11-119F7.1, CXXC6, LCX, bA119F7.1, CXXC finger 6; CXXC zinc finger 6; CXXC-type zinc finger protein 6; leukemia-associated protein with a CXXC domain; methylcytosine dioxygenase TET1; ten-eleven translocation 1 gene protein; ten-eleven translocation-1; tet oncogene 1
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80312
Immunohistochemistry (Paraffin)
OTI8G6
Unconjugated
TET1
Liquid
TET1
Human recombit protein fragment corresponding to amino acids 1-305 of TET1 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit