Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TET2 Mouse anti-Human, HRP, Clone: 649, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243552H

Catalog No. NBP243552H


Add to cart

Description

Description

TET2 Monoclonal antibody specifically detects TET2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

TET2
649
Western Blot
Purified
TET2
Recombinant protein encompassing a sequence within the N-terminus region of human TET2. The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot
HRP
PBS with No Preservative
FLJ20032, MGC125715, tet methylcytosine dioxygenase 2
Mouse
IgG1
0.1 ml
Primary
54790
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit