Learn More
TET2 Rabbit anti-Human, Alexa Fluor™ 594, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232104AF594
Catalog No. NP232104A94
Add to cart
Description
Description
TET2 Polyclonal antibody specifically detects TET2 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
TET2
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
TET2
The immunogen this antibody was made to, maps to a region between residue 100 and 150 of human Tet Oncogene Family Member 2 using the numbering given in entry NP_001120680.1 (GeneID 54790).
0.1 mL
Primary
54790
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
FLJ20032, MGC125715, tet methylcytosine dioxygenase 2
Rabbit
IgG
Store at 4°C in the dark.
Polyclonal
Human