Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TET2 Rabbit anti-Human, HRP, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232104H

Catalog No. NBP232104H


Add to cart

Description

Description

TET2 Polyclonal antibody specifically detects TET2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

TET2
HRP
PBS with No Preservative
FLJ20032, MGC125715, tet methylcytosine dioxygenase 2
Rabbit
IgG
0.1 ml
Store at 4C in the dark.
Polyclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Affinity Purified
TET2
The immunogen this antibody was made to, maps to a region between residue 100 and 150 of human Tet Oncogene Family Member 2 using the numbering given in entry NP_001120680.1 (GeneID 54790).
Immunogen affinity purified
RUO
Primary
54790
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only