Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TET3 Mouse anti-Human, Clone: GT859, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527864

Catalog No. PIMA527864


Add to cart

Description

Description

Positive Control: flag-human TET3-transfected 293T cells Predicted Reactivity: Mouse (94%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

TET3, a member of the ten-eleven-translocation (TET) family of genes, is a methylcytosine dioxygenase that catalyzes the conversion of methylcytosine to 5-hydroxymethylcytosine and is most abundantly expressed in hematopoietic cells. Unlike the related TET2 protein, mutations in TET3 have not been observed in any myeloid maligcies. TET3 has been shown to be involved in the demethylation of zygotic DNA before the first mitosis and has been suggested to be involved in the epigenetic reprogramming of the zygotic paternal DNA following natural fertilization and may also contribute to somatic cell nuclear reprogramming during animal cloning.
Specifications

Specifications

TET3
Monoclonal
1 mg/ml
PBS with no preservative
O43151
TET3, hCG_40738, tet methylcytosine dioxygenase 3
Mouse
IgG2a
100 μl
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
200424
Western Blot
GT859
Unconjugated
TET3
Liquid
TET3
Recombit protein encompassing a sequence within the C-terminus region of human TET3. The exact sequence is proprietary.
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit