Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TET3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen TET3
Applications Immunofluorescence, Immunohistochemistry
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094456
View Documents
Novus Biologicals
NBP304740100UL
100 μg Each for $320.71
Add to cart
 
NB094457
View Documents
Novus Biologicals
NBP30474020UL
20 μg Each for $138.37
Add to cart
 
Description

Description

TET3 Polyclonal antibody specifically detects TET3 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence
Specifications

Specifications

TET3
Unconjugated
Rabbit
Epigenetics
PBS with 50% glycerol, pH7.3.
200424
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence, Immunohistochemistry
Purified
RUO
Human
EC 1.14.11.n2, hCG_40738, KIAA0401, methylcytosine dioxygenase TET3, MGC22014, probable methylcytosine dioxygenase TET3, tet oncogene family member 3
Recombinant fusion protein containing a sequence corresponding to amino acids 1556-1795 of human TET3 (NP_001274420.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit