Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TET3 Rabbit anti-Mouse, DyLight 755, Polyclonal,Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP259669IR
Description
TET3 Polyclonal specifically detects TET3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TET3 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
EC 1.14.11.n2, hCG_40738, KIAA0401, methylcytosine dioxygenase TET3, MGC22014, probable methylcytosine dioxygenase TET3, tet oncogene family member 3 | |
Rabbit | |
Affinity Purified | |
Epigenetics, Histones and Modified Histones | |
200424 | |
Store at 4°C in the dark. |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TET3 | |
Partial recombinant protein made to an N-terminal portion of human TET3 (amino acids 1-216) [UniProt O43151] | |
0.1 mL | |
Primary | |
Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title