Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetanus Toxin Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10062945AF594
Description
Tetanus Toxin Polyclonal antibody specifically detects Tetanus Toxin in Bacteria samples. It is validated for ELISASpecifications
Tetanus Toxin | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Alexa Fluor 594 | |
Tentoxylysin | |
Synthetic peptide corresponding to the amino acids 1300 - 1314 of the native molecule conjugated to Keyhole limpet Haemocyanin | |
0.1 mL | |
Virology Bacteria and Parasites | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction