Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetanus Toxin Antibody, Alexa Fluor™ 647, Novus Biologicals™
Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10063239AF647
Description
Tetanus Toxin Polyclonal antibody specifically detects Tetanus Toxin in Bacteria samples. It is validated for Western Blot, ELISASpecifications
Tetanus Toxin | |
Polyclonal | |
50mM Sodium Borate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
Alexa Fluor 647 | |
Tentoxylysin | |
Purified tetanus toxin | |
0.1 mL | |
Virology Bacteria and Parasites | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction