Learn More
Tetanus toxoid Goat anti-Bacteria, Polyclonal, Abnova™
Goat Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB29819
Catalog No. 89-033-664
Add to cart
Description
Description
Goat polyclonal antibody raised against tetanus toxoid.
  • High specificity
  • High reactivity
Specifications
Specifications
Tetanus toxoid
Unconjugated
ELISA Immunofluorescence Immunohistochemistry Lateral-Flow Immunochromatographic Assay Western Blot The optimal working dilution should be determined by the end user.
Liquid
Native purified tetanus toxoid.
500 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Bacteria
ELISA, Immunofluorescence, Immunohistochemistry, Lateral Flow, Western Blot
Goat polyclonal antibody raised against tetanus toxoid.
In 0.01 M Phospate buffered saline, pH 7.2 (0.09% sodium azide).
Goat
IgG
RUO
Primary
This antibody reacts with tetanus toxoid.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)